Learn More
Description
Biotix™ Pure uTIP™ Filter Pipette Tips for Universal Pipettes, Standard Designed with Flexfit™ to lower necessary insertion and ejection force by about 70% to make secure seal. Biotix™ uTIP™ Filter Pipette Tips for Universal Pipettes are manufactured with Pure resin that is virgin polypropylene.
- Fits all standard pipettes
- FlexFit technology features flexible proximal end to ensure secure seal with 66 to 80% less insertion force than competitor tips
- Pure resin made of virgin polypropylene
- Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip
- Certified free of RNase, DNase, Pyrogens, and PCR Inhibitors
Specifications
Specifications
| Volume (Metric) | 10 μL |
| Filtered | Filtered |
| Sterility | Sterile |
| Color | Clear |
| Rack Color | Red |
| Autoclavable | Not Autoclavable |
| Length (Metric) | 45.84 mm |
| Certifications/Compliance | Certified Pre-Sterile, BioReady, Sterilized to 10-3 sterility insurance level (SAL) |
| Clean Claims | Certified Pre-Sterile, BioReady, PCR Inhibitor, Endotoxin/Pyrogen, Nucleic Acid, RNase/DNase, Sterilized to 10-3 sterility insurance level (SAL) |
| For Use With (Application) | Designed to enhance the performance and accuracy of experiments. |
| Show More |
This product contains no safety hazards
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
