Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco pure2 4 Port Left Airlock 230 V
SDP

Catalog No. 13756907 Shop All Labconco Corporation Products
Change view
Click to view available options
Voltage:
230 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
13-756-907 230 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-756-907 Supplier Labconco Supplier No. LSKPURE4GLV

Please contact your sales representative for a quote or further ordering instructions.

missing translation for 'validMainframeMsg'

The pure2 Isolator with 4-Ports and a Left Airlock (230 V) is designed for safe and efficient processing in ISO Class 5 conditions. It features advanced HEPA filtration, H2O2 decontamination, and ergonomic access.

  • Designed for Critical Processes The pure2 Isolator is engineered to support a wide range of applications requiring stringent contamination control, providing unparalleled protection for both products and users.

Unmatched Engineering for Superior Containment

  • GMP Grade A, ISO 5 Unidirectional Airflow - Maintains ultra-clean conditions, meeting Good Manufacturing Practice (GMP) and ISO 14644-1 Class 5 standards for aseptic processing.
  • Premium 316L Stainless Steel Interior - Features a smooth, seamless design that is easy to clean and resistant to corrosion, ensuring long-term durability and compliance with strict hygiene requirements.
  • Flexible Pressure Mode Selection - Supports both positive and negative pressure operation, allowing users to tailor containment conditions to their specific application needs.
  • High-Capacity, Fast-Acting Airlocks - Reduces transfer times and enhances workflow efficiency by minimizing process bottlenecks, increasing overall productivity.
  • Advanced Nanox Catalytic Converters - Enables rapid hydrogen peroxide (H2O2) decomposition, significantly shortening decontamination cycles and reducing downtime.

Precision and Reliability for Safe Operation

  • Integrated, Fast, and Safe H2O2 Decontamination - Utilizes SKANfog technology to achieve effective sterilization within each chamber, ensuring contamination-free operation.
  • High-Efficiency HEPA Filtration - Incorporates H14-grade HEPA filters for both supply and exhaust air, with dual H14 filtration on exhaust to maintain maximum environmental safety.
  • Consistent and Controlled Airflow - Provides precise air regulation to maintain validated aseptic conditions and prevent cross-contamination.

Flexible and Customizable Design

  • Modular, Space-Saving Configuration - Designed for seamless integration into diverse facility layouts, optimizing workspace utilization.
  • Multiple Glove Configurations - Available in 2-glove and 4-glove models to accommodate different process and operational requirements.
  • Extensive Customization Options - Offers a wide range of tailored features, including pass-throughs, environmental monitoring systems, and integration with specialized laboratory equipment.
  • Region-Specific Availability - The pure2 Isolator is exclusively available in the U.S., Canada, and Puerto Rico.

Specifications

Product Type Aseptic Isolator
Material Stainless Steel
Lighting Type LED
Voltage 230 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.