Learn More
CUBE BIOTECH Ni-IDA Cartridge for His-tag
Supplier: CUBE BIOTECH 30304
1ml of Ni-IDA His-tag Agarose slurry (50%) in a 1ml for Protein Purification of His-tagged proteins via FPLC
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.