Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ PureEpi™ Chromatin Preparation and Optimization Kit

Catalog No. 1710082 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
12 Chromatin Preparations
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-100-82 12 Chromatin Preparations
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-100-82 Supplier MilliporeSigma™ Supplier No. 1710082
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • 10X Glycine
  • Nuclei Isolation Buffer
  • SCW Buffer
  • Crosslink Reversal Buffer
  • Protease Inhibitor Cocktail II

Provides a complete set of reagents allowing the preparation of multiple chromatin samples from starting materials such as cultured cells or tissues

  • Complete set of materials for up to 12 chromatin preparations
  • Optimized streamlined protocol with only a single buffer for sonication, IP, or wash
  • Protocols for using cells or tissues
  • Compatible with Magna ChIP HiSens and EZ-Magna ChIP HiSens chromatin immunoprecipitation kits
  • Compatible with ChIPAb+ validated antibody and primer sets
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store components at the temperatures indicated on the labels
For Use With (Application) Preparation of multiple chromatin samples from starting materials such as cultured cells or tissues
Quantity 12 Chromatin Preparations
Stability Kit components are stable for 1 year from date of shipment when stored as directed
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.