Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo 3000 NIOSH-Approved HEPA Filters
SDP

Catalog No. 18444006
Change view
Click to view available options
Quantity:
25/Pk.
5/Pk.
50/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-444-006 50/Pk.
18-444-004 5/Pk.
18-444-005 25/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 18-444-006 Supplier Gentex™ Supplier No. PF300002002MP
Add to Cart
Add to Cart
This item is not returnable. View return policy

NIOSH-approved HEPA Filters for use with the PureFlo 3000 PAPR system.

  • NIOSH-approved HEPA Filters
  • For use with the PureFlo 3000 PAPR system
  • Replaceable

Specifications

Certifications (NIOSH) NIOSH Approved
Color Gray
Filter Type HEPA
Product Type Filter
Filter Efficiency High Efficiency
For Use With (Equipment) PureFlo 3000 PAPR Systems
Latex Free Yes
Model PF3000-02-002MP
Quantity 50/Pk.
Replaceable Filter Yes
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.