Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo ESM+ PF60 PAPR Respirator System, Black Hard Hat
SDP

Catalog No. 18444060
Change view
Click to view available options
Includes:
Black Hard Hat, Black Stedprene FR Neck Cape, HE Filter, APF 1000, Internal Switch
Black Hard Hat, Black Stedprene FR Neck Cape, HE Filter, APF 1000, Internal and External Switches
Black Hard Hat, Black Stedprene FR Neck Cape, HE/HF/HC Filter, APF 1000, Internal Switch
Black Hard Hat, Black Stedprene FR Neck Cape, HE/HF/HC Filter, APF 1000, Internal and External Switches
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
18-444-060 Black Hard Hat, Black Stedprene FR Neck Cape, HE Filter, APF 1000, Internal Switch
18-444-059 Black Hard Hat, Black Stedprene FR Neck Cape, HE Filter, APF 1000, Internal and External Switches
18-444-064 Black Hard Hat, Black Stedprene FR Neck Cape, HE/HF/HC Filter, APF 1000, Internal Switch
18-444-066 Black Hard Hat, Black Stedprene FR Neck Cape, HE/HF/HC Filter, APF 1000, Internal and External Switches
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 18-444-060 Supplier Gentex™ Supplier No. PR05801B0A1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Self-contained PAPRs are ideal for medium to heavy-duty applications. Innovative all-in-one design provides head, eye, face, and respiratory protection against hazardous airborne contaminants, sparks, flying debris, chemical and molten metal splashes.

  • Includes ANSI-approved hard hat, HEPA filter, battery pack, visor assembly, and cape
  • Charger sold separately
  • Electronic Safety Management System (ESM+) includes LED status indicator and audible alarm for low flow, low battery, and filter change
  • Adjustable flame retardant neck cape
  • No cables, hoses, or belt-mounted blower
  • Unique continuous airflow system with airflow rate of 8.1 cfm
  • Wide vision ANSI approved polycarbonate face shield
  • Quick-release headband for easy adjustment and donning
  • HEPA (HE) Filter (other filters available)
  • Corrosion-resistant gold plated battery
  • Choose from internal and external on/off switch or internal switch only

Specifications

ANSI Standard Head Protection: ANSI Z89.1 Type 1 Class G; Face Shield/Eye Protection ANSI Z87.1+: 2015 Certified; SEI Certified and Listed
Assigned Protection Factor (APF) 1000
Battery Life 7 hours with two batteries
Battery Type 5 Cell Nickel Metal Hydride
Cartridge/Filter PARTICULATE
Certifications (NIOSH) NIOSH Approved
Product Type PAPR Hard Hat System
Size One Size
Filter Efficiency HEPA (HE) or HEPA/HF/HC *Nuisance OV
Flow Rate 8.1 CFM or 230 L/min.
Includes Black Hard Hat, Black Stedprene FR Neck Cape, HE Filter, APF 1000, Internal Switch
Model PR05801-B-0A1
Noise Level Under 70 dB
Quantity 1 Ea.
Recharge Time 2.5 hr.
Replaceable Filter One Size
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.