Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo ESM+ Power Source Replacement Parts and Accessories
SDP

Catalog No. 18444047
Change view
Click to view available options
Product Type:
Battery Charger
Battery Pack
Multi-Unit Charging Station
Color:
Black
Gray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Color
18-444-047 Battery Charger Black
18-444-046 Battery Pack Black
18-444-048 Multi-Unit Charging Station Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 18-444-047 Supplier Gentex™ Supplier No. PR02040SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Replacement parts including battery, charger, and multi-charger for use with PureFlo ESM+ PAPR systems.

  • Power source replacement parts and accessories for the PureFlo ESM+ system include
    • Battery pack
    • Battery charger
    • Battery multicharger - 16 bank

Specifications

Color Black
Disposable Reusable
Material Plastic
Product Type Battery Charger
Size One Size
For Use With (Equipment) PureFlo ESM+ PAPR
Latex Free Yes
Model PR02040SP
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.