Learn More
Purell™ Advanced Hand Sanitizer Gel: Refill for NXT™ Dispenser
Shop All Gojo Industries, Inc. Products

Description
- Kills of most common germs that may cause illness
- Maintains skin health
- Refill pack options feature removable pump for easier recycling and are SANITARY SEALED™ to prevent contamination
Specifications
Specifications
| Product Type | Hand Sanitizer Refill |
| Color | Clear |
| Form | Gel |
| For Use With (Equipment) | Refill for PURELL™ NXT™ Dispenser |
| Quantity | 4 Pk. |
| Volume (Metric) | 1000 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.