Learn More
Description
- Provides a comprehensive solution to help reduce the risk of Norovirus outbreaks
- Safely removes vomit, blood, feces & other body fluids
- Effective against Norovirus, E.Coli, Salmonella and more in just 30 seconds
- Proven effective on most hard and soft surfaces
- Protects staff and patrons from exposure to biohazards
- Bilingual, step-by-step instructions for ease of use
- OSHA and Food Code compliant
- Approved for food contact surfaces (no rinse required)
- Includes hairnet, sleeved gown, gloves, shoe covers, eye shield and mask to provide head-to-toe personal protection
- 3-year shelf life
Compliance
OSHA and Food Code Compliant
Specifications
Specifications
| Product Type | Body Fluid Spill Kit |
| For Use With (Application) | Body fluid spills |
| Length (English) | 27.5 in |
| Length (Metric) | 69.85 cm |
| Color | White |
| Material | Plastic |
| Sterility | Non-sterile |
| Packaging | White Plastic Bag |
| Quantity | 16/Pk. |
| Dimensions (L x W) | 27.5 x 17.5 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
