Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PURELL™ Healthcare Advanced Hand Sanitizer Foam
Shop All Gojo Industries, Inc. Products

Click to view available options
For Use With (Equipment):
ES4 dispensing system
ES6 dispensing system
ES8 dispensing system
Description
Advanced Hand Sanitizer Foam formulation designed for healthcare environments. AT-A-GLANCE refills make it obvious when refills need to be changed.

Specifications
Specifications
Product Type | Hand Sanitizer |
Color | Clear |
Form | Foam |
For Use With (Equipment) | ES6 dispensing system |
Size | 1200 mL |
Volume (Metric) | 1200 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction