Learn More
Description
- Convenient, easy-to-use and ideal for offices, restaurants, health clubs—anywhere
- Textured for superior cleaning
- Sanitize hands while wiping away light soils and dirt
- Kill 99.99% of most common germs that may cause illness
Specifications
Specifications
| Product Type | Sanitizing Wipe |
| Certifications/Compliance | TSCA,15 USC 2602 (2)(B), FDA, WHMIS |
| For Use With (Application) | For superior cleaning wipes away light soil |
| Packaging Quantity | 4000 Individually Packed |
| Size | 4000 Sheets/Cs. |
| pH Range | 4.5 to 9.5 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
