Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purell™ NXT™ Space Saver™ Dispenser
SDP

Catalog No. 19043730 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Color:
Dove Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-043-730 Dove Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-043-730 Supplier Gojo Industries Inc Supplier No. 212006

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

This item is not returnable. View return policy

Much smaller footprint than traditional 800mL dispenser

Ideal for applications with limited wall space or where decor or usage levels demand smaller dispensers.

  • Uses 1000mL NXT refills
  • Compact design integrates into most every decor

Compliance

Fully ADA depth and push compliant

TRUSTED_SUSTAINABILITY

Specifications

Product Type NXT Space Saver Dispenser
Color Dove Gray
Certifications/Compliance ADA
For Use With (Application) Limited wall space or where decor or usage levels demand smaller dispensers
For Use With (Equipment) 1000mL NXT refills
Includes Nonclog valve
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.