Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purell™ Professional Advanced Hand Sanitizer Foam 5054-02
SDP

Catalog No. 19043862 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Volume (Metric):
1200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19-043-862 1200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-043-862 Supplier Gojo Industries Inc Supplier No. 505402
This item is not returnable. View return policy

Enhanced with unique skin conditioners to maintain skin health. Purell™ Professional Advanced Hand Sanitizer Foam 5054-02 kills more than 99.99% of most common germs that may cause illness.

  • Advanced foam hand sanitizer formulation for professional environments
  • Kills more than 99.99% of most common germs that may cause illness
  • Outperforms other hand sanitizers ounce for ounce
  • America's #1 brand of instant hand sanitizer
  • Enhanced with unique skin conditioners to maintain skin health
  • Hypoallergenic
  • Dermatologist tested
  • Dye Free
  • Energy-on-the-refill technology - every refill has a built-in energy source, designed to last as long as the refill
  • AT-A-GLANCE™ refills make it obvious when refills need to be changed
  • SANITARY SEALED™ PET refill with the tear-off collar for easy recyclability
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Sanitizer
Color Clear
For Use With (Equipment) ES4 Push-Style Hand Sanitizer Dispensers
Quantity 2 Pk.
Volume (Metric) 1200 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.