Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purell™ Professional Advanced Hand Sanitizer Fragrance Free Foam 6452-02
SDP

Catalog No. 19043860 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Volume (Metric):
1200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19-043-860 1200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-043-860 Supplier Gojo Industries Inc Supplier No. 645202

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Enhanced with unique skin conditioners to maintain skin health. Purell™ Professional Advanced Hand Sanitizer Fragrance Free Foam 6452-02 kills more than 99.99% of most common germs that may cause illness.

  • Advanced foam hand sanitizer formulation for professional environments
  • Kills more than 99.99% of most common germs that may cause illness
  • Outperforms other hand sanitizers ounce for ounce
  • America's #1 brand of instant hand sanitizer
  • Enhanced with unique skin conditioners to maintain skin health
  • Hypoallergenic
  • Dermatologist tested
  • Dye Free
  • Energy-on-the-refill technology - every refill has a built-in energy source, designed to last as long as the refill
  • AT-A-GLANCE™ refills make it obvious when refills need to be changed
  • SANITARY SEALED™ PET refill with the tear-off collar for easy recyclability
  • ECOLOGO™ Certified to UL 2783 Hand Sanitizer Standard
  • USDA Biobased Certified
  • Clinically proven formulation to maintain skin health
  • Supports LEED certification criteria
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Sanitizer
Color Clear
For Use With (Equipment) ES6 Touch-Free Hand Sanitizer Dispensers
Quantity 2 Pk.
Volume (Metric) 1200 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.