Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purell™ Professional HEALTHY SOAP™ Mild Foam
SDP

Catalog No. 19043830 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
PURELL ES6 Touch-Free Soap Dispensers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-043-830 PURELL ES6 Touch-Free Soap Dispensers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-043-830 Supplier Gojo Industries Inc Supplier No. 647402

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

PURELL™ Professional HEALTHY SOAP™ Mild Foam for PURELL™ Soap Dispensers.

  • Exceptionally mild
  • Fragrance-free
  • Green-certified foam hand soap
  • Washes away dirt and germs
  • Enriched with a moisturizer and skin conditioners
  • Contains natural moisturizers
  • Nourishes the skin while cleaning
  • Supports your skin's natural ability to protect itself against dirt and germs
  • Hypoallergenic
  • AT-A-GLANCE™ refills make it obvious when refills need to be changed
  • SANITARY SEALED™ PET refill is easily recycled
TRUSTED_SUSTAINABILITY

Specifications

Product Type Foam Hand Wash
Color Blue
Certifications/Compliance Meets UL EcoLogo Standard for Hand Cleaner UL 2784, Meets USDA biobased Standard for hand cleaners
For Use With (Application) Hand Washing
For Use With (Equipment) PURELL ES6 Touch-Free Soap Dispensers
Quantity 2 Pk.
Volume (English) 40.58 oz.
Volume (Metric) 1200 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.