Learn More
MilliporeSigma™ Supelco™ Purge/Trap K Vocarb™ 3000
Efficiently traps analytes in US EPA Methods 502.2 and 524
Supplier: MilliporeSigma™ Supelco™ 24940U
Description
- Tubes used in purge and trap analyses generally are packed with multiple beds of adsorbent materials
- A single tube can trap a broad range of polar and nonpolar, high and low molecular weight compounds
- Each bed protects the next, increasingly active bed, by preventing compounds from being held so strongly that they cannot be desorbed quickly without decomposition
- During the purge phase of sampling, lower molecular weight compounds pass through the initial adsorbent beds but are trapped by succeeding beds
- During desorption, the carrier gas passes through the trap in the reverse direction of purge flow, so that higher molecular weight compounds never come in contact with the stronger (innermost) adsorbents
Specifications
Purge/Trap K | |
Industrial Hygiene | |
OI Analytical Eclipse 4660, 4560 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vocarb is a registered trademark of Siemens Industry, Inc.
Spot an opportunity for improvement?Share a Content Correction