Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ PuriCare™ Procedure Station Class II, Type A2 Biosafety Cabinet, US Model
SDP

Catalog No. 01812537 Shop All Labconco Corporation Products
Change view
Click to view available options
Includes:
Cabinet
Cabinet, Accessory Package
Plug Type:
North America (115 V, 15 A)
North America (115 V, 20 A)
North America (230 V)
Width (English) Exterior:
42.4 in.
54.4 in.
66.4 in.
78.4 in.
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Plug Type Width (English) Exterior
01-812-537 Cabinet North America (115 V, 20 A) 66.4 in.
01-812-545 Cabinet North America (115 V, 15 A) 42.4 in.
01-812-543 Cabinet, Accessory Package North America (115 V, 15 A) 42.4 in.
01-812-541 Cabinet North America (115 V, 15 A) 54.4 in.
01-812-539 Cabinet, Accessory Package North America (115 V, 15 A) 54.4 in.
01-812-535 Cabinet, Accessory Package North America (115 V, 20 A) 66.4 in.
01-812-533 Cabinet North America (115 V, 20 A) 78.4 in.
01-812-531 Cabinet, Accessory Package North America (115 V, 20 A) 78.4 in.
01-812-544 Cabinet North America (230 V) 42.4 in.
01-812-542 Cabinet, Accessory Package North America (230 V) 42.4 in.
01-812-540 Cabinet North America (230 V) 54.4 in.
01-812-538 Cabinet, Accessory Package North America (230 V) 54.4 in.
01-812-536 Cabinet North America (230 V) 66.4 in.
01-812-534 Cabinet, Accessory Package North America (230 V) 66.4 in.
01-812-532 Cabinet North America (230 V) 78.4 in.
01-812-530 Cabinet, Accessory Package North America (230 V) 78.4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 01-812-537 Supplier Labconco™ Supplier No. 312520002
Only null left
Add to Cart
Add to Cart
Request a Quote

PuriCare Procedure Stations create a particulate-free environment for small animal research activities including surgical procedures, cage changing, weighing, and anesthesia delivery. Models feature a 12 inch sash opening and washable prefilter.

Performance Features:

  • Safe Class II airflows (approx. 70% recirculating, 30% exhausted) that provide personnel, product, and environmental protection.
    • Cabinet interior provides ISO Class 5 (Grade A) or better working conditions.
  • Constant Airflow Profile™ (CAP) and BalanceFlow™ technology maintain safe cabinet airflows – even as HEPA filters load.
    • Optimized airflow minimizes energy use and noise output to industry-leading levels.
  • Removable, washable prefilters to protect HEPA longevity.
  • Logic OS with 5 in. line-of-sight touchscreen display.
    • Access cabinet information such as filter life, alerts and alarms, data logs.
    • Remote connectivity, particle counter integration, and certification records elevate user control of the cabinet.
  • Smart-Start™ and Night-Smart™ modes simplify startup and shut-down procedures to help ensure consistency and cleanliness before, during, and after work.
  • Inclination Technology™ improves ergonomic working conditions by maximizing comfort and accessibility during long periods of work.
  • Mag-Zone™ area and mating accessories keep common items such as pipettes and biohazard waste organized and within reach.
  • Full, six-year warranty on all Class II biosafety cabinets.

Optional Configurations:

  • Accessory Package: UV lamp (254 nm), right-side service fixture with ball-type valve and right and left-side Vacu-Pass™ Cord and Cable Portals

Specifications

Description 5 ft. PuriCare Procedure Station Class II, Type A2 Biosafety Cabinet
Certifications/Compliance NSF/ANSI 49, ETL, UL, CAN/CSA, ADA, ISO 5 Conditions per ISO 14644-1 and 2
Chamber Material Stainless Steel (304)
Width (Metric) Interior 153.7 cm
Product Type Class II, Type A2 Bio Safety Cabinet
Model PuriCare Procedure Station
Noise Level 66 dBA
Outlets 2 Duplex
Plug Type North America (115 V, 20 A)
Power Consumption 340 W
Receptacles 2 Double, Left and Right Side
Height (English) 61.9 in.
Sash Opening (Metric) 30.5 cm
Width (English) Exterior 66.4 in.
Volume (Metric) Exhaust 1196 m³/hr.
Display 5 in. Touchscreen
Filter Efficiency 99.99%
Filter Type HEPA
Width (Metric) Exterior 168.7 cm
Includes Cabinet
Energy Consumption (Reduced Flow Mode) 50 W
Lighting LED
Height (Metric) 157.2 cm
Sash Opening (English) 12 in.
Service Fixtures 0
Volume 704 CFM
Warranty 6 Years
Width (English) Interior 60.5 in.
Frequency 60 Hz
Height (English) Interior 25.7 in.
Height (English) Max. Opening 22.5 in.
Height (English) Viewing 27 in.
Height (Metric) Interior 65.3 cm
Height (Metric) Max. Opening 57.2 cm
Height (Metric) Viewing 69 cm
Depth (English) 32 in.
Depth (Metric) 81.3 cm
Voltage 115 V
Energy Consumption (Operating Set Point) 340 W
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.