Learn More
Biolegend Purified anti-E. coli RNA Polymerase α
Supplier:  Biolegend 663104
Encompass_Preferred
Purified anti-E. coli RNA Polymerase α [4RA2]; Isotype: Mouse IgG1; Reactivity: E. coli, Widely among Gram-negative bacteria, some Gram-positive bacteria; Apps: WB, ELISA, IP; Size: 100 ug
Catalog No. 50-208-7201