Learn More
Biolegend Purified anti-HIV gp120
Supplier:  Biolegend 906801
Encompass_Preferred
Purified anti-HIV gp120 [ED8.D4]; Isotype: Mouse IgG1; Reactivity: HIV-1; Apps: WB, ELISA; Size: 250 uL
Catalog No. 50-207-5172
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.