Learn More
Biolegend Purified anti-human CD360 (IL-21R)
Supplier:  Biolegend 347802
Encompass_Preferred
Purified anti-human CD360 (IL-21R) [2G1-K12]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 ug
Catalog No. NC1633424
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.