Learn More
Biolegend Purified anti-Myosin Heavy Cha
Supplier:  Biolegend 922703
Encompass_Preferred
Purified anti-Myosin Heavy Chain [H11]; Isotype: Mouse IgM, κ; Reactivity: Human; Apps: WB, ICC; Size: 25 μg
Catalog No. 50-272-1618
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.