Learn More
Biolegend Purified anti-SIRT7
Supplier:  Biolegend 666101
Encompass_Preferred
Purified anti-SIRT7 [W17121A]; Isotype: Rat IgG2a, κ; Reactivity: Human, Mouse; Apps: WB, IP; Size: 25 ug
Catalog No. 50-208-7236