Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Purified HMGB2 (NP_002120.1, 1 a.a. - 209 a.a.) human Recombinant Protein with His-Flag-StrepII tag at N-terminus expressed in human cells

Catalog No. 89953483 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-483 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-953-483 Supplier Abnova™ Supplier No. H00003148H01
Add to Cart
Add to Cart

Used for ELISA, PI, SDS-PAGE, WB

This gene encodes a member of the non-histone chromosomal high mobility group protein family. The proteins of this family are chromatin-associated and ubiquitously distributed in the nucleus of higher eukaryotic cells. In vitro studies have demonstrated that this protein is able to efficiently bend DNA and form DNA circles. These studies suggest a role in facilitating cooperative interactions between cis-acting proteins by promoting DNA flexibility. This protein was also reported to be involved in the final ligation step in DNA end-joining processes of DNA double-strand breaks repair and V(D)J recombination. [provided by RefSeq]

Sequence: MGKGDPNKPRGKMSSYAFFVQTCREEHKKKHPDSSVNFAEFSKKCSERWKTMSAKEKSKFEDMAKSDKARYDREMKNYVPPKGDKKGKKKDPNAPKRPPSAFFLFCSEHRPKIKSEHPGLSIGDTAKKLGEMWSEQSAKDKQPYEQKAAKLKEKYEKDIAAYRAKGKSEAGKKGPGRPTGSKKKNEPEDEEEEEEEEDEDEEEEDEDEE

Specifications

Accession Number NP_002120.1
Concentration ≥ 10 ug/ml
For Use With (Application) ELISA, Protein Interaction, SDS-PAGE, Western Blot
Formulation Liquid
Gene ID (Entrez) 3148
Molecular Weight (g/mol) 29.28kDa
Name HMGB2 (Human) Recombinant Protein
Preparation Method Transfection of pSuper-HMGB2 plasmid into HEK293T cell, and the expressed protein was purified by Strep-Tactin affinity column.
Purification Method Strep-Tactin affinity columns
Quality Control Testing SDS-PAGE and Western Blot
Quantity 2 μg
Immunogen MGKGDPNKPRGKMSSYAFFVQTCREEHKKKHPDSSVNFAEFSKKCSERWKTMSAKEKSKFEDMAKSDKARYDREMKNYVPPKGDKKGKKKDPNAPKRPPSAFFLFCSEHRPKIKSEHPGLSIGDTAKKLGEMWSEQSAKDKQPYEQKAAKLKEKYEKDIAAYRAKGKSEAGKKGPGRPTGSKKKNEPEDEEEEEEEEDEDEEEEDEDEE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HMG2
Common Name HMGB2
Gene Symbol HMGB2
Biological Activity Not Tested
Species Human
Recombinant Recombinant
Protein Tag His-Flag-StrepII
Expression System Transfection of pSuper-HMGB2 plasmid into HEK293T cell, and the expressed protein was purified by Strep-Tactin affinity column.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.