Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Purified LAMP3 (AAH32940.1 28 a.a. - 381 a.a.) human Recombinant Protein with His-Flag-StrepII tag at N-terminus expressed in human cells

Catalog No. 89953486 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-486 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-953-486 Supplier Abnova™ Supplier No. H00027074H01
Add to Cart
Add to Cart

Used for ELISA, PI, SDS-PAGE, WB

Sequence: KAFPGTRDYSQPTAAATVQDIKKPVQQPAKQAPHQTLAARFMDGHITFQTAATVKIPTTTPATTKNTATTSPITYTLVTTQATPNNSHTAPPVTEVTVGPSLAPYSLPPTITPPAHTTGTSSSTVSHTTGNTTQPSNQTTLPATLSIALHKSTTGQKPVQPTHAPGTTAAAHNTTRTAAPASTVPGPTLAPQPSSVKTGIYQVLNGSRLCIKAEMGIQLIVQDKESVFSPRRYFNIDPNATQASGNCGTRKSNLLLNFQGGFVNLTFTKDEESYYISEVGAYLTVSDPETVYQGIKHAVVMFQTAVGHSFKCVSEQSLQLSAHLQVKTTDVQLQAFDFEDDHFGNVDECSSDYT

Specifications

Accession Number AAH32940.1
Concentration ≥ 10 ug/ml
For Use With (Application) ELISA, Protein Interaction, SDS-PAGE, Western Blot
Formulation Liquid
Gene ID (Entrez) 27074
Molecular Weight (g/mol) 44.22kDa
Name LAMP3 (Human) Recombinant Protein
Preparation Method Transfection of pSuper-LAMP3 plasmid into HEK293T cell, and the expressed protein was purified by Strep-Tactin affinity column.
Purification Method Strep-Tactin affinity columns
Quality Control Testing SDS-PAGE and Western Blot
Quantity 25 μg
Immunogen KAFPGTRDYSQPTAAATVQDIKKPVQQPAKQAPHQTLAARFMDGHITFQTAATVKIPTTTPATTKNTATTSPITYTLVTTQATPNNSHTAPPVTEVTVGPSLAPYSLPPTITPPAHTTGTSSSTVSHTTGNTTQPSNQTTLPATLSIALHKSTTGQKPVQPTHAPGTTAAAHNTTRTAAPASTVPGPTLAPQPSSVKTGIYQVLNGSRLCIKAEMGIQLIVQDKESVFSPRRYFNIDPNATQASGNCGTRKSNLLLNFQGGFVNLTFTKDEESYYISEVGAYLTVSDPETVYQGIKHAVVMFQTAVGHSFKCVSEQSLQLSAHLQVKTTDVQLQAFDFEDDHFGNVDECSSDYT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CD208/DC-LAMP/DCLAMP/LAMP/TSC403
Common Name LAMP3
Gene Symbol LAMP3
Biological Activity Not Tested
Species Human
Recombinant Recombinant
Protein Tag His-Flag-StrepII
Expression System Transfection of pSuper-LAMP3 plasmid into HEK293T cell, and the expressed protein was purified by Strep-Tactin affinity column.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.