Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Purified TNFRSF1A (AAH10140.1 22 a.a. - 211 a.a.) human Recombinant Protein with His-Flag-StrepII tag at N-terminus expressed in human cells (H02)

Catalog No. 89953503 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-503 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-953-503 Supplier Abnova™ Supplier No. H00007132H02

Used for ELISA, PI, SDS-PAGE, WB

The protein encoded by this gene is a member of the TNF-receptor superfamily. This protein is one of the major receptors for the tumor necrosis factor-alpha. This receptor can activate NF-kappaB, mediate apoptosis, and function as a regulator of inflammation. Antiapoptotic protein BCL2-associated athanogene 4 (BAG4/SODD) and adaptor proteins TRADD and TRAF2 have been shown to interact with this receptor, and thus play regulatory roles in the signal transduction mediated by the receptor. Germline mutations of the extracellular domains of this receptor were found to be associated with the autosomal dominant periodic fever syndrome. The impaired receptor clearance is thought to be a mechanism of the disease. [provided by RefSeq]

Sequence: IYPSGVIGLVPHLGDREKRDSVCPQGKYIHPQNNSICCTKCHKGTYLYNDCPGPGQDTDCRECESGSFTASENHLRHCLSCSKCRKEMGQVEISSCTVDRDTVCGCRKNQYRHYWSENLFQCFNCSLCLNGTVHLSCQEKQNTVCTCHAGFFLRENECVSCSNCKKSLECTKLCLPQIENVKGTEDSGTT

Specifications

Accession Number AAH10140.1
Concentration ≥ 10 ug/ml
For Use With (Application) ELISA, Protein Interaction, SDS-PAGE, Western Blot
Formulation Liquid
Gene ID (Entrez) 7132
Molecular Weight (g/mol) 26.18kDa
Name TNFRSF1A (Human) Recombinant Protein
Preparation Method Transfection of pSuper-TNFRSF1A plasmid into HEK293H cell, and the expressed protein was purified by Strep-Tactin affinity column.
Purification Method Strep-Tactin affinity columns
Quality Control Testing SDS-PAGE and Western Blot
Quantity 2 μg
Immunogen IYPSGVIGLVPHLGDREKRDSVCPQGKYIHPQNNSICCTKCHKGTYLYNDCPGPGQDTDCRECESGSFTASENHLRHCLSCSKCRKEMGQVEISSCTVDRDTVCGCRKNQYRHYWSENLFQCFNCSLCLNGTVHLSCQEKQNTVCTCHAGFFLRENECVSCSNCKKSLECTKLCLPQIENVKGTEDSGTT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CD120a/FPF/MGC19588/TBP1/TNF-R/TNF-R-I/TNF-R55/TNFAR/TNFR1/TNFR55/TNFR60/p55/p55-R/p60
Common Name TNFRSF1A
Gene Symbol TNFRSF1A
Biological Activity Not Tested
Species Human
Recombinant Recombinant
Protein Tag His-Flag-StrepII
Expression System Transfection of pSuper-TNFRSF1A plasmid into HEK293H cell, and the expressed protein was purified by Strep-Tactin affinity column.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.