Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Purified TNFRSF9 (AAH06196.1 24 a.a. - 186 a.a.) human Recombinant Protein with His-Flag-StrepII tag at N-terminus expressed in human cells

Catalog No. 89953506 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-506 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-953-506 Supplier Abnova™ Supplier No. H00003604H03
Add to Cart
Add to Cart

Used for ELISA, PI, SDS-PAGE, WB

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor contributes to the clonal expansion, survival, and development of T cells. It can also induce proliferation in peripheral monocytes, enhance T cell apoptosis induced by TCR/CD3 triggered activation, and regulate CD28 co-stimulation to promote Th1 cell responses. The expression of this receptor is induced by lymphocyte activation. TRAF adaptor proteins have been shown to bind to this receptor and transduce the signals leading to activation of NF-kappaB. [provided by RefSeq]

Sequence: LQDPCSNCPAGTFCDNNRNQICSPCPPNSFSSAGGQRTCDICRQCKGVFRTRKECSSTSNAECDCTPGFHCLGAGCSMCEQDCKQGQELTKKGCKDCCFGTFNDQKRGICRPWTNCSLDGKSVLVNGTKERDVVCGPSPADLSPGASSVTPPAPAREPGHSPQ

Specifications

Accession Number AAH06196.1
Concentration ≥ 10 ug/ml
For Use With (Application) ELISA, Protein Interaction, SDS-PAGE, Western Blot
Formulation Liquid
Gene ID (Entrez) 3604
Molecular Weight (g/mol) 23.21kDa
Name TNFRSF9 (Human) Recombinant Protein
Preparation Method Transfection of pSuper-TNFRSF9 plasmid into HEK293H cell, and the expressed protein was purified by Strep-Tactin affinity column.
Purification Method Strep-Tactin affinity columns
Quality Control Testing SDS-PAGE and Western Blot
Quantity 25 μg
Immunogen LQDPCSNCPAGTFCDNNRNQICSPCPPNSFSSAGGQRTCDICRQCKGVFRTRKECSSTSNAECDCTPGFHCLGAGCSMCEQDCKQGQELTKKGCKDCCFGTFNDQKRGICRPWTNCSLDGKSVLVNGTKERDVVCGPSPADLSPGASSVTPPAPAREPGHSPQ
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias 4-1BB/CD137/CDw137/ILA/MGC2172
Common Name TNFRSF9
Gene Symbol TNFRSF9
Biological Activity Not Tested
Species Human
Recombinant Recombinant
Protein Tag His-Flag-StrepII
Expression System Transfection of pSuper-TNFRSF9 plasmid into HEK293H cell, and the expressed protein was purified by Strep-Tactin affinity column.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.