Learn More
ZeptoMetrix Purified Viral Lysate, Metapneumovirus 3 Type B1
Shop All ZeptoMetrix Corporation Products

Description
- Available in 1mg vials
IMPORTANT: Contains Metapneumovirus. Before vendor can ship order, customer must submit a Certificate of Compliance (COC) regarding use restrictions.
Specifications
Specifications
| Biosafety Level | 2 |
| Certifications/Compliance | ISO, 13485 |
| Concentration | 0.5 to 3mg/mL |
| Content And Storage | -65°C |
| Detects | Viral Antibodies |
| Form | Solution |
| Media Type | Vial |
| Product Type | Purified Viral Lysate |
| For Use With (Application) | Dot blotting, Immunodetection of Antibodies to Adenovirus using solid phase Enzyme immunoassays, western blotting, Other Protein based assay |
| Purification Method | Sucrose density gradient ultracentrifugation |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.