Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puritan™ PurFlock™ Ultra Non-Sterile Flocked Swabs

Catalog No. 22029520
Change view
Click to view available options
Length (Metric):
150.6 mm
152.4 mm
152.9 mm
153.2 mm
153.4 mm
Tip Style:
Elongated Flocked
Large
Micro Ultrafine
Minitip flocked
Regular flocked
Ultrafine Flocked
Length (Metric) Handle:
100 mm
80 mm
80 or 100 mm
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Tip Style Length (Metric) Handle
22-029-520 152.4 mm Elongated Flocked 80 or 100 mm
22-029-605 150.6 mm Large 80 mm
22-029-609 153.2 mm Micro Ultrafine 100 mm
22-029-518 152.4 mm Elongated Flocked 100 mm
22-029-606 153.4 mm Regular flocked 80 mm
22-029-607 153.4 mm Minitip flocked 80 or 100 mm
22-029-608 152.9 mm Ultrafine Flocked 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 22-029-520 Supplier Puritan™ Supplier No. 3806U
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ideal for various rapid diagnostic tests

PurFlock's purity make it ideal for PCR, molecular assays, DFA testing, forensic applications as well as direct antigen testing. PurFlock Ultra’s high purity characteristics makes it an ideal swab for various rapid diagnostic tests.

Certifications

CE marked (for medical use), ISO 9001:2008, ISO 13485:2003

Compliance

FDA Good Manufcturing Practices (GMP)

Specifications

Certifications/Compliance CE, ISO 9001:2008, ISO 13485:2003, FDA GMP's
Length (English) 6 in.
For Use With (Application) Collection of Microorganisms, Buccal cells and forensic evidence
Product Type Flocked Swab
Length (Metric) 152.4 mm
Length (English) Handle 3.14 or 4 in.
Length (Metric) Handle 80 or 100 mm
Length (English) Tip 0.685 in.
Length (Metric) Tip 17.39 mm
Diameter (Metric) Tip 5.5 mm
Diameter (English) Tip 0.219 in.
Packaging Bag
Sterility Non-sterile
Handle Material Polystyrene
Tip Style Elongated Flocked
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.