Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puromycin Dihydrochloride,, MP Biomedicals

Catalog No. ICN10055290 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
250 mg
500 mg
1 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10055290 500 mg
ICN10055210 10 mg
ICN10055225 25 mg
ICN10055280 100 mg
ICN10055283 250 mg
ICN10055291 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. ICN10055290 Supplier MP Biomedicals, Inc Supplier No. 0210055290
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58-58-2
Molecular Formula C22H31Cl2N7O5
Molecular Weight (g/mol) 544.43
MDL Number MFCD00150080
InChI Key MKSVFGKWZLUTTO-MLYJQVMKNA-N
Synonym Stylomycin, 6-dimethylamino-9-(3'-deoxy-3'-(p-methoxy-L-phenylalanyamino)- B-D-ribofuranosyl)-purine
PubChem CID 131632508
SMILES Cl.Cl.COC1=CC=C(C[C@H](N)C(=O)N[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O)N2C=NC3=C(N=CN=C23)N(C)C)C=C1

Specifications

Type Puromycin Dihydrochloride
Physical Form Powder
Extinction Coefficient 20.3 (275nm, 0.1N NaOH), 19.5 (268nm, 0.1 NHCl) (Lit.)
Color White
Quantity 500 mg
IUPAC Name (2S)-2-amino-N-[(2S,3S,4S,5R)-5-[6-(dimethylamino)purin-9-yl]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]-3-(4-methoxyphenyl)propanamide;dihydrochloride
Formula Weight 544.4
Chemical Name or Material Puromycin Dihydrochloride, from Streptomyces albo-niger
EINECSNumber 200-387-8
Recommended Storage Store at - 0°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.