Learn More
Description
- Applications-Immunohistochemistry, Immunohistochemistry-Paraffin
- Format-Ready-to-use
- Storage-Room temperature
- Protocol-
- Rinse slides with deionized or distilled water.
- Place slides into Purple Hematoxylin Solution for 30 to 60 seconds.
- Rinse slides in deionized or distilled water until all free hematoxylin has been removed.
- Bluing in an alkaline solution is not required.
Specifications
Specifications
| For Use With (Application) | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Product Line | Novus |
| Quantity | 100 mL |
| Storage Requirements | Room temperature |
| Technique | Preparation of Working Solutions Purple Hematoxylin is ready to use and does not require any preparation. Protocol Recommendations Counterstain 1. Rinse slides with deionized or distilled water. 2. Place slides into Purple Hematoxylin for 30-60 seconds. 3. Rinse slides in deionized or distilled water until all free hematoxylin has been removed. 4. Bluing the slides in an alkaline solution is not required and is not recommended. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
