Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PUS7 Monoclonal Antibody (OTI4A2), TrueMAB™, OriGene
SDP

Catalog No. p-200010534
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1832 30 μL
50-167-3135 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1832 Supplier OriGene Supplier No. TA502049S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PUS7 Monoclonal antibody specifically detects PUS7 in Canine, Human, Rat samples. It is validated for Western Blot

Specifications

Antigen PUS7
Applications Western Blot
Classification Monoclonal
Clone OTI4A2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ANXA3
Gene Accession No. P12429, P14669
Gene Alias C330017I15Rik; FLJ20485; Kiaa1897; pseudouridine synthase 7; pseudouridylate synthase 7; pseudouridylate synthase 7 (putative); pseudouridylate synthase 7 homolog; pseudouridylate synthase 7 homolog (S. cerevisiae); PUS7; RGD1307054
Host Species Mouse
Immunogen Full length human recombinant protein of human ANXA3 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25291, 306, 478447
Target Species Canine, Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.