Learn More
MilliporeSigma™ Supelco™ Push Button Valve
Catalog No. 111011948 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
VICI Series C and D Pressure-Lok Syringes
Catalog No. For Use With (Equipment)
11-101-1948 VICI Series C and D Pressure-Lok Syringes
1 options
Catalog No. 11-101-1948 Supplier MilliporeSigma™ Supelco™ Supplier No. 22284
Add to Cart
Add to Cart
This Mininert valve will attach to your syringe and provide sample storing capability. It features push-button operation and an all-PTFE body with a stainless steel miniature valve stem.
This Mininert valve will attach to your syringe and provide sample storing capability. It features push-button operation and an all-PTFE body with a stainless steel miniature valve stem.
TRUSTED_SUSTAINABILITY
Specifications
Material PTFE
Quantity 1/Pk.
For Use With (Equipment) VICI Series C and D Pressure-Lok Syringes