Learn More
AVANTOR MASTERFLEX (VWR LLC) 2-Stop, PVC Solvent/Hydrocarbon Tubing, 1.30 mm ID, 12/pk
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX9560632
Ismatec SC0164 2-Stop, PVC Solvent/Hydrocarbon Tubing, 1.30 mm ID, 12/pk, Designed for optimal operation in your Ismatec pumpsIdeal for chromatography applications and for the transfer of fuels, distillates, lubricants, and glycol-based coolantsResistant to ozone and UV light; resistant to cracking, swelling, and hardeningAppearance: Yellow, translucentTemperature: ¿30 to 85¿C