Learn More
Agilent Technologies PVPACK-GP VECTOR
Supplier:  Agilent Technologies 217566
Encompass
pVPack-GP Vector is a gag-pol-expressing vector that is cotransfected with the retroviral expression vector together with one of a choice of 4 envelope (env)-expressing vector.
Catalog No. 50-125-293
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.