Learn More
AdipoGen PX-12 (5 mg)
SDP
Supplier:  AdipoGen AGCR13739M005
Encompass_Preferred
Chemical. CAS: 141400-58-0. Formula: C7H12N2S2. MW: 188.3. PX-12 is a competitive, irreversible inhibitor of thioredoxin 1 (Trx1), acting by binding to Cys73 of this enzyme. PX-12 stimulates apoptosis, down-regulates HIF-1alpha, vascular endothelial growth factor (VEGF) and iNOS, and consequently inhibits proliferation and tumor growth in animal models. It has also been reported to inhibit tubulin polymerization in vitro, induce cell cycle arrest at G2/M phase and inhibit growth, migration and invasion of several cancer cell lines. PX-12 shows antiviral activity against HIV-1, by inhibiting the enzymatic activity of Trx1 and the Trx1-dependent disulfide reduction of gp120. Microtubule polymerization and formation of acetylated microtubules are inhibited as well, activities shown to be required for HIV-1 life cycle propagation. PX-12 potentially inhibits the Mpro/3C-like protease of SARS-CoV-2, consequently inhibiting viral transcription and replication.
Catalog No. 50-205-4168
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.