Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPB™ PX5™ PAPR Power Source Replacement Parts and Accessories
SDP

Power sources include rechargeable battery and complete charger kit designed for the PX5™ powered air-purifying respirator system.

Specifications

Color Gray
Disposable Reusable
Material Plastic
Size Universal
For Use With (Equipment) PX5™ PAPR
View More Specs

Products 1
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
17-000-013
SDP
View Documents
RPB™
03855
Battery Pack N/A N/A N/A
Description

Description

  • Rechargeable lithium-ion battery and complete charger kit
  • Replacement Parts for PX5™
Specifications

Specifications

Gray
Plastic
PX5™ PAPR
Reusable
Universal
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.