Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PXDN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5144080
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5144080 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5144080 Supplier Invitrogen™ Supplier No. PA5144080
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control - WB: human Hela whole cell, human U251 whole cell, rat heart tissue, mouse heart tissue. IHC: human pancrease cancer tissue, human renal cancer tissue.

Drosophila peroxidasin is an extracellular matrix-associated peroxidase (Horikoshi et al., 1999 [PubMed 10441517]). It is expressed exclusively in hemocytes derived from head mesoderm at a very early stage of differentiation. Peroxidasin exists as a homotrimer with a unique hybrid structure that combines an enzymatically functional peroxidase domain with motifs that are typically found in extracellular matrix-associated proteins. It is a secreted protein that contains a secretory recognition sequence at its N terminus. Peroxidasin catalyzes hydrogen peroxide-driven radioiodination, oxidations, and the formation of dityrosine in vitro. It is also thought to function in extracellular matrix consolidation, phagocytosis, and defense.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PXDN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene PXDN
Gene Accession No. Q3UQ28, Q92626
Gene Alias 2310075M15Rik; C85409; COPOA; D2S448; D2S448E; E330004E07; Kiaa0230; Melanoma-associated antigen MG50; MG50; mKIAA0230; p53-responsive gene 2 protein; PE responsive protein c64; Perc64; peroxidasin; peroxidasin homolog; peroxidasin homolog (Drosophila); PRG2; PXDN; PXN; Vascular peroxidase 1; VPO; VPO1
Gene Symbols PXDN
Host Species Rabbit
Immunogen Human PXDN between 900-1064.
Purification Method Antigen affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 554172, 69675, 7837
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.