Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PXMP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501734551 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4551 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4551 Supplier Proteintech Group Inc Supplier No. 248011AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Peroxisomes are subcellular organelles with important functions in lipid metabolism that are found in virtually all eucaryotic cells. The peroxisomal membrane contains a number of integral and peripheral membrane proteins involved in the import of peroxisomal matrix proteins and the transport of metabolites across the membrane. Pxmp2 (Peroxisomal membrane protein 2), also known as PMP22, is a 195 amino acid multi-pass membrane protein that belongs to the peroxisomal membrane protein PXMP2/4 family. The Pxmp2 protein interacts with both Peroxin 19 and Siva proteins. The Peroxin 19 protein binds multiple peroxisomal membrane proteins (PMPs), is predomitly cytoplasmic, and is required for peroxisome membrane synthesis. Pxmp2 is the most abundant peroxisomal membrane protein in higher eukaryotes. Pxmp2′s expression is tissue-specific with highest levels of expression in liver, kidney and heart tissue. It has been suggested that Pxmp2 is involved in pore-forming activity and may contribute to the broad permeability of the peroxisomal membrane. Disruption of the mouse Pxmp2 gene leads to partial restriction of peroxisomal membrane permeability to solutes in vitro and in vivo. The Pxmp2 gene is conserved in chimpanzee, dog, cow, mouse, rat, zebrafish, fruit fly and mosquito, and maps to human chromosome 12q24.33.

Specifications

Antigen PXMP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PXMP2
Gene Accession No. P42925, Q07066, Q9NR77
Gene Alias Peroxisomal membrane protein 2, PMP22, PXMP2
Gene Symbols PXMP2
Host Species Rabbit
Immunogen PXMP2 Fusion Protein Ag20590
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19301, 29533, 5827
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.