Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PXR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541170
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541170 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541170 Supplier Invitrogen™ Supplier No. PA541170
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KKEMIMSDEA VEERRALIKR KKSERTGTQP LGVQGLTEEQ RMMIRELMDA Sequence homology: Human: 100%.

This gene product belongs to the nuclear receptor superfamily, members of which are transcription factors characterized by a ligand-binding domain and a DNA-binding domain. The encoded protein is a transcriptional regulator of the cytochrome P450 gene CYP3A4, binding to the response element of the CYP3A4 promoter as a heterodimer with the 9-cis retinoic acid receptor RXR. It is activated by a range of compounds that induce CYP3A4, including dexamethasone and rifampicin. Several alternatively spliced transcripts encoding different isoforms, some of which use non-AUG translation initiation codon, have been described for this gene. Additional transcript variants exist, however, they have not been fully characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PXR
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NR1I2
Gene Accession No. O75469
Gene Alias BXR; mPXR; Nr1i2; Nuclear receptor subfamily 1 group I member 2; nuclear receptor subfamily 1, group 1, member 2; nuclear receptor subfamily 1, group I, member 2; ONR 1; ONR1; orphan nuclear receptor PAR1; Orphan nuclear receptor PXR; PAR; PAR 1; PAR 2; PAR q; PAR1; PAR2; PARq; pregnane X nuclear receptor variant 2; Pregnane X receptor; pregnane X receptor (nuclear receptor sub family 1, group I, member 2); PRR; PXR; PXR.1; PXR.2; PXR1; SAR; Steroid and xenobiotic receptor; SXR
Gene Symbols NR1I2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human NR1I2 (aa 108-157).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8856
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.