Learn More
Medchemexpress LLC HY-D1106 5mg Medchemexpress, Py-BODIPY-NHS ester CAS:201998-61-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD11065MG
Encompass_Preferred
Medchemexpress, HY-D1106 5mg Py-BODIPY-NHS ester CAS:201998-61-0 Py-BODIPY-NHS ester is a cell-permeable and fluorescent energy transfer probe. Py-BODIPY-NHS ester tracer yields strong cellular engagement and proves to be optimal for MET kinase. The live-cell energy transfer technique can perform a broad-coverage, quantitative analysis of intracellular kinase target engagement. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2753
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.