Learn More
Anaerobe Systems PYG BROTH - Peptone Yeast Extract Broth with Glucose, 10 Tubes/Pkg
Supplier: Anaerobe Systems AS822
This item has a minimum qty of 2 per supplier requirements.
Peptone Yeast Extract Broth Based Media are enriched nonselective broth used for growth & identification of anaerobic bacteria based on their biochemical features. This media is prepared, dispensed, & packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use.PY Broth contains hemin & vitamin K for the recovery of fastidious organisms such as Prevotella spp & Porphyromonas spp. PRAS (Pre-reduced anaerobically sterilized) biochemicals are considered the gold standard for biochemical testing & characterization of anaerobes. Adding specific biochemical, the Peptone Yeast Extract Broth Based Media can determine whether anaerobic bacteria will utilize the biochemical or if growth is stimulated or inhibited. PY was originally formulated for use in chromatographic analysis of metabolic products from the fermentation of glucose.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.