Learn More
Abcam Pyocyanin, Catalase Inhibitor, >98% Purity, Solid, 25 mg
Description
- MW: 210.23 Da
- Purity: >98%
- Form: Solid
- Activity: Catalase inhibitor, aryl hydrocarbon receptor activator (Ki = 5.4 μM)
- Biological effects: Reduces catalase gene expression, promotes extracellular DNA release, generates ROS, and induces apoptosis in neutrophils in vivo
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Catalase, Aryl Hydrocarbon Receptor |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Substrate Properties | Chemical Substrate |
| Quantity | 25 mg |
| For Use With (Application) | Catalase inhibition, Aryl hydrocarbon receptor activation, ROS generation, Apoptosis induction |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.