Learn More
Apexbio Technology LLC PYR-41 418805-02-4 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B14925.1
Encompass_Preferred
PYR-41 (CAS 418805-02-4) is a selective inhibitor of ubiquitin-activating enzyme (E1) involved in the ubiquitination cascade essential for diverse cellular processes such as protein degradation apoptosis DNA repair and signaling pathways By inhibiting E1 activity PYR-41 impedes ubiquitination reactions resulting in decreased proteasomal degradation In vitro studies demonstrate that PYR-41 also increases overall cellular SUMOylation and attenuates cytokine-induced NF- B activation through interference with TRAF6-dependent (non-proteasomal) ubiquitination and stabilization of I B Currently PYR-41 remains in preclinical research serving primarily as an investigative tool to explore ubiquitination mechanisms and related signal transduction pathways
Catalog No. 50-000-62000
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.