Learn More
AdipoGen Pyraflufen-ethyl
SDP
Supplier:  AdipoGen CDXP0169M025
Encompass_Preferred
Chemical. CAS 129630-19-9. Formula C15H13Cl2F3N2O4. MW 413.18. Synthetic. Phenylpyrazole herbicide derivative of pyraflufen. Used to control broad-leaved weeds and grasses in crops. Action of mode is through inhibition of protoporphyrinogen oxidase PPO.
Catalog No. 50-256-0211
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.