Learn More
New Pig Pyramex™ Class 2 Cooling Safety Vest
SDP
Catalog No. 17987945 Shop All New Pig Corporation Products
Catalog No. 17-987-945 Supplier New Pig Supplier No. WPL7372X5XNG
Add to Cart
Add to Cart
Work comfortably for an 8 hour shift in this special material cooling vest with adjustable waistband and soft outer shell
  • Extended Cooling Relief: The vest's evaporative cooling technology provides up to 8 hours of cooling comfort to help workers manage heat stress during a full shift.
  • Specialized Cooling Material: Made with an inner evaporative PVA material that activates when soaked in cool water for 1-2 minutes, delivering long-lasting relief.
  • Adjustable and Comfortable Fit: Features an adjustable waistband and a soft quilted outer shell, allowing for a comfortable and customized fit for any wearer.
  • High-Visibility Safety: Meets ANSI/ISEA 107-2015 Class 2 standards for high-visibility apparel, ensuring workers are easily seen in high-traffic or low-light areas.
  • Durable and Reusable: Constructed for durability, this cooling vest can be washed and reused, making it a cost-effective and sustainable solution for managing heat.
  • Convenient Design: The vest includes a zipper front for easy on and off, and is made with high-visibility lime polyester and reflective striping to enhance safety.