Description
- Durable dual injected polycarbonate and rubber temples
- Single lens design provides maximum protection
- Lenses are coated for superior scratch resistance
- Lenses provide 99% protection against harmful UV rays
Specifications
Specifications
| Lens Tint | Clear |
| Frame Color | Gray |
| Lens Material | Polycarbonate |