Learn More
Description
- Universal sizing fits on the brim of most hard hats and accommodates a variety of face shields
- Note: Model HHAN (19-151-614) is dielectric
Specifications
Specifications
| Certifications/Compliance | ANSI Z87.1 |
| Color | Blue |
| For Use With (Equipment) | Cap-style Hard Hats and Variety of Faceshields |
| Material | Nylon |
| Product Type | Hard Hat Adapter |
| Size | Universal |
| Style | Dielectric |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.