Description
Ratcheting lens pitch adjustment aligns lens angle to properly position for work application.
- Flexible nylon temples with four size settings to accommodate a broad range of facial sizes
- Adjustable nose pad conforms to a broad range of bridge sizes
- Lightweight nylon frame for comfortable, all day wear
- Replaceable, 9.5 base-curved polycarbonate lens withstands high-impact
- Provide 99% UV protection, is completely distortion free
Compliance
Meet ANSI Z87.1 requirements.
Specifications
Specifications
| Certifications/Compliance | Meet ANSI Z87.1 requirements. |
| Lens Tint | Infinity Blue |
| Frame Color | Black |
| Lens Material | Polycarbonate |
| Frame Material | Polycarbonate |