Learn More
Pyramex™ Rendezvous™ Safety Glasses
Catalog No. 19121007 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Black
Blue
Crystal Gray
Lens Tint:
Amber
Clear
Gray
Indoor/Outdoor Mirror
Infinity Blue
Silver Mirror
Lens Coating:
Scratch-resistant
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint Lens Coating
19-121-007 Black Silver Mirror -
19-121-004 Black Clear Scratch-resistant
19-152-340 Black Clear Scratch-resistant
19-121-005 Black Gray -
19-152-343 Black Amber -
19-121-009 Black Indoor/Outdoor Mirror Scratch-resistant
19-152-341 Crystal Gray Clear Scratch-resistant
19-121-006 Black Infinity Blue -
19-121-008 Black Gray -
19-121-010 Blue Clear -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 19-121-007 Supplier Pyramex™ Supplier No. SB2870S
This item is not returnable. View return policy
Adjustable lens angle aligns to the proper position for various work applications
Ratcheting lens pitch adjustment aligns lens angle to properly position for work application.
  • Flexible nylon temples with four size settings to accommodate a broad range of facial sizes
  • Adjustable nose pad conforms to a broad range of bridge sizes
  • Lightweight nylon frame for comfortable, all day wear
  • Replaceable, 9.5 base-curved polycarbonate lens withstands high-impact
  • Provide 99% UV protection, is completely distortion free
Compliance
Meet ANSI Z87.1 requirements.
Specifications
Certifications/Compliance Meet ANSI Z87.1 requirements.
Lens Tint Silver Mirror
Frame Color Black
Temple Style Flexible
Lens Material Polycarbonate
Frame Material Polycarbonate