Learn More
Pyramex™ SX3 Plus™ Safety Glasses
Catalog No. 19817390 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
Mirror, Indoor/Outdoor
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-817-390 Clear
19-817-415 Gray
19-155-738 Mirror, Indoor/Outdoor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-817-390 Supplier Pyramex™ Supplier No. SB4610STP
Add to Cart
Add to Cart
This item is not returnable. View return policy
Sports-look safety spectacle with superior eye protection.
  • Open temples allow clip on cord option for goggle-like fit
  • Unique slim temple design with co-injection temple tips
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection.
  • Soft foam padding provides comfort fit; elastic strap
Specifications
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Safety Glasses
For Use With (Application) Eye protection
Frame Color Black
Temple Style Adjustable
Lens Material Polycarbonate